(S)-tert-Butyl 2-amino-3-methylbutylcarbamate 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR452193-1G OR452193-250MG OR452193-500MG
APOSOR452193-1GEA 1060 EUR
APOSOR452193-1G APOSOR452193-250MG APOSOR452193-500MG
(S)-tert-Butyl 2-amino-3-methylbutylcarbamate 97%
(S)-tert-Butyl 2-amino-3-methylbutylcarbamate
Formule: C₁₀H₂₂N₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD18694786
CAS nummer: 400652-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR