2-(4-Chlorophenoxy)-benzylamine 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR452224-1G OR452224-500MG
APOSOR452224-1GEA 526 EUR
APOSOR452224-1G APOSOR452224-500MG
2-(4-Chlorophenoxy)-benzylamine 97%
2-(4-Chlorophenoxy)-benzylamine
Formule: C₁₃H₁₂ClNO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11191890
CAS nummer: 792158-57-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR