1H-Pyrazolo[3,4-b]pyridine-3-carboxaldehyde 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR460010-1G
APOSOR460010-1GEA 1020 EUR
APOSOR460010-1G
1H-Pyrazolo[3,4-b]pyridine-3-carboxaldehyde 97%
1H-Pyrazolo[3,4-b]pyridine-3-carboxaldehyde
Formule: C₇H₅N₃O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11100578
CAS nummer: 1010073-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR