(R)-(+)-4-Bromo-α-methylbenzyl alcohol

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR460032-5G OR460032-1G OR460032-250MG
APOSOR460032-5GEA 519 EUR
APOSOR460032-5G APOSOR460032-1G APOSOR460032-250MG
(R)-(+)-4-Bromo-α-methylbenzyl alcohol
(R)-(+)-4-Bromo-α-methylbenzyl alcohol
Formule: C₈H₉BrO
Dichtheid: 1,322 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06201861
CAS nummer: 76155-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR