3-Bromo-5-iodophenol

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR46047-1G OR46047-250MG OR46047-5G
APOSOR46047-1GEA 24.9 EUR
APOSOR46047-1G APOSOR46047-250MG APOSOR46047-5G
3-Bromo-5-iodophenol
3-Bromo-5-iodophenol
Formule: C₆H₄BrIO
Smeltpunt: 82…84 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD12405397
CAS nummer: 570391-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR