tert-Butyl-5-oxo-D-prolinate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR46095-25G
APOSOR46095-25GEA 64.3 EUR
APOSOR46095-25G
tert-Butyl-5-oxo-D-prolinate
tert-Butyl-5-oxo-D-prolinate
Formule: C₉H₁₅NO₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09261417
CAS nummer: 205524-46-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR