4-(Methylsulphonyl)benzyl bromide 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR46201-10G OR46201-5G OR46201-100G OR46201-25G OR46201-1G
APOSOR46201-10GEA 0 EUR
APOSOR46201-10G APOSOR46201-5G APOSOR46201-100G APOSOR46201-25G APOSOR46201-1G
4-(Methylsulphonyl)benzyl bromide 97%
4-(Methylsulphonyl)benzyl bromide
Formule: C₈H₉BrO₂S
Smeltpunt: 82…86 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00185849
CAS nummer: 53606-06-7
UN: 3261
ADR: 8,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR