2,5-Dimethoxythiophenol

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR4657-1G
APOSOR4657-1GEA 35.1 EUR
APOSOR4657-1G
2,5-Dimethoxythiophenol
2,5-Dimethoxythiophenol
Formule: C₈H₁₀O₂S
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00052738
CAS nummer: 1483-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR