2-Bromo-3-chlorobenzenesulphonamide

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR46616-1G OR46616-250MG OR46616-5G
APOSOR46616-1GEA 93.5 EUR
APOSOR46616-1G APOSOR46616-250MG APOSOR46616-5G
2-Bromo-3-chlorobenzenesulphonamide
2-Bromo-3-chlorobenzenesulphonamide
Formule: C₆H₅BrClNO₂S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD18390303
CAS nummer: 1261499-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR