2-Oxoglutaric acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR4662-100G OR4662-25G OR4662-500G
APOSOR4662-100GEA 67.7 EUR
APOSOR4662-100G APOSOR4662-25G APOSOR4662-500G
2-Oxoglutaric acid
2-Oxoglutaric acid
Formule: HOOCCH₂CH₂COCOOH
Smeltpunt: 113…117 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00004165
CAS nummer: 328-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR