N-Boc-Pro-Gly-OMe

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR470084-1G OR470084-5G
APOSOR470084-1GEA 257 EUR
APOSOR470084-1G APOSOR470084-5G
N-Boc-Pro-Gly-OMe
N-Boc-Pro-Gly-OMe
Formule: C₁₃H₂₂N₂O₅
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD16495870
CAS nummer: 34290-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR