5-Hydroxy-2-(methylsulphonamido)benzoic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR470319-1G OR470319-5G
APOSOR470319-1GEA 277 EUR
APOSOR470319-1G APOSOR470319-5G
5-Hydroxy-2-(methylsulphonamido)benzoic acid
5-Hydroxy-2-(methylsulphonamido)benzoic acid
Formule: C₈H₉NO₅S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD14660388
CAS nummer: 1243391-74-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR