4-(Methylamino)piperidine dihydrochloride

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR470425-25G OR470425-5G
APOSOR470425-25GEA 446 EUR
APOSOR470425-25G APOSOR470425-5G
4-(Methylamino)piperidine dihydrochloride
4-(Methylamino)piperidine dihydrochloride
Formule: C₆H₁₄N₂·2ClH
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04039797
CAS nummer: 1220039-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR