Methyl 4-Aminopiperidine-4-carboxylate dihydrochloride

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR470509-1G OR470509-5G
APOSOR470509-1GEA 400 EUR
APOSOR470509-1G APOSOR470509-5G
Methyl 4-Aminopiperidine-4-carboxylate dihydrochloride
Methyl 4-Aminopiperidine-4-carboxylate dihydrochloride
Formule: C₇H₁₄N₂O₂·2ClH
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09998913
CAS nummer: 161315-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR