5-Bromo-7-chlorobenzofuran

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR470778-1G OR470778-250MG
APOSOR470778-1GEA 1020 EUR
APOSOR470778-1G APOSOR470778-250MG
5-Bromo-7-chlorobenzofuran
5-Bromo-7-chlorobenzofuran
Formule: C₈H₄BrClO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11877832
CAS nummer: 1194375-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR