4-(Bromomethyl)quinoline

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR471097-1G OR471097-250MG
APOSOR471097-1GEA 358 EUR
APOSOR471097-1G APOSOR471097-250MG
4-(Bromomethyl)quinoline
4-(Bromomethyl)quinoline
Formule: C₁₀H₈BrN
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09953523
CAS nummer: 5632-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR