5-(Bromomethyl)-2-(methylthio)pyrimidine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR471324-1G
APOSOR471324-1GEA 646 EUR
APOSOR471324-1G
5-(Bromomethyl)-2-(methylthio)pyrimidine
5-(Bromomethyl)-2-(methylthio)pyrimidine
Formule: C₆H₇BrN₂S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD24465277
CAS nummer: 323591-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR