2-(Methylthio)-4,5-pyrimidinedicarboxylic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR471456-250MG OR471456-1G OR471456-5G
APOSOR471456-250MGEA 206 EUR
APOSOR471456-250MG APOSOR471456-1G APOSOR471456-5G
2-(Methylthio)-4,5-pyrimidinedicarboxylic acid
2-(Methylthio)-4,5-pyrimidinedicarboxylic acid
Formule: C₇H₆N₂O₄S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD13195439
CAS nummer: 149771-16-4

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR