2-(5-Ethyl-1,3,4-oxadiazol-2-yl)ethylamine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR471579-1G
APOSOR471579-1GEA 1550 EUR
APOSOR471579-1G
2-(5-Ethyl-1,3,4-oxadiazol-2-yl)ethylamine
2-(5-Ethyl-1,3,4-oxadiazol-2-yl)ethylamine
Formule: C₆H₁₁N₃O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09883756
CAS nummer: 944898-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR