2-(1H-Pyrrol-2-yl)acetonitrile

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR471598-1G OR471598-250MG OR471598-5G
APOSOR471598-1GEA 409 EUR
APOSOR471598-1G APOSOR471598-250MG APOSOR471598-5G
2-(1H-Pyrrol-2-yl)acetonitrile
2-(1H-Pyrrol-2-yl)acetonitrile
Formule: C₆H₆N₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09878875
CAS nummer: 50551-29-6

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR