4-Amino-2-(tert-butyl)pyrimidine 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR471697-1G OR471697-250MG
APOSOR471697-1GEA 729 EUR
APOSOR471697-1G APOSOR471697-250MG
4-Amino-2-(tert-butyl)pyrimidine 95%
4-Amino-2-(tert-butyl)pyrimidine
Formule: C₈H₁₃N₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01646228
CAS nummer: 114362-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR