1-(4-Bromophenyl)-N-methylethanamine hydrochloride

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR475087-1G OR475087-250MG
APOSOR475087-1GEA 132 EUR
APOSOR475087-1G APOSOR475087-250MG
1-(4-Bromophenyl)-N-methylethanamine hydrochloride
1-(4-Bromophenyl)-N-methylethanamine hydrochloride
Formule: C₉H₁₂BrN·ClH
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD31695722
CAS nummer: 799293-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR