5-Chloronicotinic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR4770-1G OR4770-5G
APOSOR4770-1GEA 22 EUR
APOSOR4770-1G APOSOR4770-5G
5-Chloronicotinic acid
5-Chloronicotinic acid
Formule: C₆H₄ClNO₂
Smeltpunt: 169…171 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00128901
CAS nummer: 22620-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR