1,3-Bis(2,6-diisopropylphenyl)imidazolinium chloride 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR48001-1G OR48001-5G OR48001-100MG OR48001-10G
APOSOR48001-1GEA 22 EUR
APOSOR48001-1G APOSOR48001-5G APOSOR48001-100MG APOSOR48001-10G
1,3-Bis(2,6-diisopropylphenyl)imidazolinium chloride 95%
1,3-Bis(2,6-diisopropylphenyl)imidazolinium chloride
Formule: C₂₇H₃₉ClN₂
Smeltpunt: 289…293 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD07369796
CAS nummer: 258278-25-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR