1-(7-Bromonaphthalen-1-yl)cyclopropane-1-carboxylic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR480144-1G
APOSOR480144-1GEA 4570 EUR
APOSOR480144-1G
1-(7-Bromonaphthalen-1-yl)cyclopropane-1-carboxylic acid
1-(7-Bromonaphthalen-1-yl)cyclopropane-1-carboxylic acid
Formule: C₁₄H₁₁BrO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD30496584
CAS nummer: 1781387-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR