4,6-Dihydro-1H-pyrrolo[3,4-c]pyrazole-5-carboxylic acid tert-butyl ester 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR480248-1G OR480248-250MG OR480248-500MG
APOSOR480248-1GEA 469 EUR
APOSOR480248-1G APOSOR480248-250MG APOSOR480248-500MG
4,6-Dihydro-1H-pyrrolo[3,4-c]pyrazole-5-carboxylic acid tert-butyl ester 95%
4,6-Dihydro-1H-pyrrolo[3,4-c]pyrazole-5-carboxylic acid tert-butyl ester
Formule: C₁₀H₁₅N₃O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08061257
CAS nummer: 657428-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR