(3-Methoxyphenyl)hydrazine hydrochloride 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR480591-100G
APOSOR480591-100GEA 175 EUR
APOSOR480591-100G
(3-Methoxyphenyl)hydrazine hydrochloride 98%
(3-Methoxyphenyl)hydrazine hydrochloride
Formule: C₇H₁₁ClN₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00044416
CAS nummer: 39232-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR