3,5-Dibromothiophene-2-carboxylic acid 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR480644-5G
APOSOR480644-5GEA 506 EUR
APOSOR480644-5G
3,5-Dibromothiophene-2-carboxylic acid 95%
3,5-Dibromothiophene-2-carboxylic acid
Formule: C₅H₂Br₂O₂S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01006744
CAS nummer: 7311-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR