2-Isocyanoethylbenzene 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR480865-1G OR480865-5G
APOSOR480865-1GEA 813 EUR
APOSOR480865-1G APOSOR480865-5G
2-Isocyanoethylbenzene 95%
2-Isocyanoethylbenzene
Formule: C₉H₉N
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD02664670
CAS nummer: 59795-89-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR