2-Bromo-4-iodo-1-isopropoxybenzene 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR48087-1G OR48087-250MG
APOSOR48087-1GEA 658 EUR
APOSOR48087-1G APOSOR48087-250MG
2-Bromo-4-iodo-1-isopropoxybenzene 95%
2-Bromo-4-iodo-1-isopropoxybenzene
Formule: C₉H₁₀BrIO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD28129290
CAS nummer: 1369903-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR