4-Bromopyridazine hydrobromide 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR49036-1G OR49036-250MG OR49036-5G
APOSOR49036-1GEA 217 EUR
APOSOR49036-1G APOSOR49036-250MG APOSOR49036-5G
4-Bromopyridazine hydrobromide 95%
4-Bromopyridazine hydrobromide
Formule: C₄H₃BrN₂·BrH
MW: 239.896 g/mol
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD15474999
CAS nummer: 1220039-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR