(Pentamethylcyclopentadienyl)iridium(III) dichloride dimer

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Dichloro(pentamethylcyclopentadienyl)iridium(III) Dimer, Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer, [Ir(Cp-)Cl2]2, (Pentamethylcyclopentadienyl)iridium (III) chloride dimer

OR49087-1G
APOSOR49087-1GEA 284 EUR
APOSOR49087-1G
(Pentamethylcyclopentadienyl)iridium(III) dichloride dimer
(Pentamethylcyclopentadienyl)iridium(III) dichloride dimer
Formule: C₂₀H₃₀Cl₄Ir₂
MW: 796.69 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00075435
CAS nummer: 12354-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR