4-Chlorobenzyl alcohol 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: (4-Chlorophenyl)methanol

OR5022-100G OR5022-25G OR5022-500G
APOSOR5022-100GEA 0 EUR
APOSOR5022-100G APOSOR5022-25G APOSOR5022-500G
4-Chlorobenzyl alcohol 98%
4-Chlorobenzyl alcohol
Formule: C₇H₇ClO
MW: 142.58 g/mol
Kookpunt: 234 °C
Smeltpunt: 68…71 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004652
CAS nummer: 873-76-7
EINECS: 212-852-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR