5-Bromo-2,4-dimethylbenzoic acid 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR50986-1G OR50986-250MG
APOSOR50986-1GEA 424 EUR
APOSOR50986-1G APOSOR50986-250MG
5-Bromo-2,4-dimethylbenzoic acid 99%
5-Bromo-2,4-dimethylbenzoic acid
Formule: C₉H₉BrO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD24107212
CAS nummer: 842136-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR