Ethyl 5-hydroxynicotinate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 3-(Ethoxycarbonyl)-5-hydroxypyridine, Ethyl 5-hydroxypyridine-3-carboxylate

OR51770-1G OR51770-250MG
APOSOR51770-1GEA 43.8 EUR
APOSOR51770-1G APOSOR51770-250MG
Ethyl 5-hydroxynicotinate
Ethyl 5-hydroxynicotinate
Formule: C₈H₉NO₃
MW: 167.162 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00510027
CAS nummer: 59288-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR