2-Iodo-6-methylbenzyl alcohol 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR51813-1G OR51813-250MG OR51813-5G
APOSOR51813-1GEA 290 EUR
APOSOR51813-1G APOSOR51813-250MG APOSOR51813-5G
2-Iodo-6-methylbenzyl alcohol 95%
2-Iodo-6-methylbenzyl alcohol
Formule: C₈H₉IO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD18397812
CAS nummer: 1261794-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR