2,4-Dihydroxy-6-methylbenzaldehyde

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR51894-1G OR51894-5G OR51894-10G
APOSOR51894-1GEA 26.3 EUR
APOSOR51894-1G APOSOR51894-5G APOSOR51894-10G
2,4-Dihydroxy-6-methylbenzaldehyde
2,4-Dihydroxy-6-methylbenzaldehyde
Formule: C₈H₈O₃
MW: 152.147 g/mol
Smeltpunt: 180…184 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00033852
CAS nummer: 487-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR