5-Thiazolamine hydrochloride

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR51952-1G
APOSOR51952-1GEA 114 EUR
APOSOR51952-1G
5-Thiazolamine hydrochloride
5-Thiazolamine hydrochloride
Formule: C3H5ClN2S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD10687116
CAS nummer: 942631-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR