Methyl 4,5,6,7-tetrahydrobenzo[b]thiophene-2-carboxylate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

OR52032-10G OR52032-1G OR52032-5G
APOSOR52032-10GEA 804 EUR
APOSOR52032-10G APOSOR52032-1G APOSOR52032-5G
Methyl 4,5,6,7-tetrahydrobenzo[b]thiophene-2-carboxylate
Methyl 4,5,6,7-tetrahydrobenzo[b]thiophene-2-carboxylate
Formule: C₁₀H₁₂O₂S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06654963
CAS nummer: 91489-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR