3-Bromo-5-nitrobenzaldehyde 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR52145-10G OR52145-1G OR52145-5G
APOSOR52145-10GEA 131 EUR
APOSOR52145-10G APOSOR52145-1G APOSOR52145-5G
3-Bromo-5-nitrobenzaldehyde 95%
3-Bromo-5-nitrobenzaldehyde
Formule: C₇H₄BrNO₃
Smeltpunt: 98…100 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00100097
CAS nummer: 355134-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR