Methyl 4-amino-2,5-dimethylbenzoate 95+%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR52228-5G
APOSOR52228-5GEA 213 EUR
APOSOR52228-5G
Methyl 4-amino-2,5-dimethylbenzoate 95+%
Methyl 4-amino-2,5-dimethylbenzoate
Formule: C₁₀H₁₃NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09839220
CAS nummer: 21339-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR