2-Chloorfenol 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 1-Chloro-2-hydroxybenzene

OR52255-100G OR52255-25G OR52255-1G OR52255-5G
APOSOR52255-100GEA 48.3 EUR
APOSOR52255-100G APOSOR52255-25G APOSOR52255-1G APOSOR52255-5G
2-Chloorfenol 99%
2-Chloorfenol

Has been used as a reagent for the protection of OH-groups in oligonucleotide synthesis. A study of various cleavage methods showed that the tetramethylguanidine salts of either 4-nitrobenzaldoxime or pyridine-2-carboxaldoxime gave the best results: Tetrahedron Lett., 2727 (1978).

Formule: C₆H₅ClO
MW: 128.56 g/mol
Kookpunt: 175…176 °C
Smeltpunt: 7…8 °C
Dichtheid: 1.26 g/cm³ (25 °C)
Vlampunt: 64 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002159
CAS nummer: 95-57-8
EINECS: 202-433-2
UN: 2021
ADR: 6.1 III
Merck Index: 12,02173

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR