Bis[(pinacolato)boryl]methane 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR52256-1G OR52256-5G OR52256-25G
APOSOR52256-1GEA 26.3 EUR
APOSOR52256-1G APOSOR52256-5G APOSOR52256-25G
Bis[(pinacolato)boryl]methane 95%
Bis[(pinacolato)boryl]methane

Uses: A Useful Methylene Unit Usable for Suzuki-Miyaura Cross-coupling ;One-Pot Synthesis of Symmetrical and Unsymmetrical Diarylmethanes; ?-Alkylation of bis[(pinacolato)boryl]methane and its application to the asymmetric Suzuki-Miyaura cross-coupling

Formule: C₁₃H₂₆B₂O₄
Smeltpunt: 53 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD27977747
CAS nummer: 78782-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR