Boc-b-Alanine 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR52389-25G OR52389-500G OR52389-100G OR52389-1KG
APOSOR52389-25GEA 0 EUR
APOSOR52389-25G APOSOR52389-500G APOSOR52389-100G APOSOR52389-1KG
Boc-b-Alanine 98%
Boc-b-Alanine
Formule: C₈H₁₅NO₄
Kookpunt: 138…142 °C/0,3mmHg
Smeltpunt: 76…78 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00037291
CAS nummer: 3303-84-2
EINECS: 221-979-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR