2-Bromo-4-chloropyrimidine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR52503-1G OR52503-100MG OR52503-250MG
APOSOR52503-1GEA 366 EUR
APOSOR52503-1G APOSOR52503-100MG APOSOR52503-250MG
2-Bromo-4-chloropyrimidine
2-Bromo-4-chloropyrimidine
Formule: C₄H₂BrClN₂
Smeltpunt: 47…48 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11847634
CAS nummer: 885702-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR