3-Bromo-2-nitrobenzyl alcohol 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR52700-1G OR52700-250MG OR52700-5G
APOSOR52700-1GEA 291 EUR
APOSOR52700-1G APOSOR52700-250MG APOSOR52700-5G
3-Bromo-2-nitrobenzyl alcohol 95%
3-Bromo-2-nitrobenzyl alcohol
Formule: C₇H₆BrNO₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD18392284
CAS nummer: 1261475-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR