(S)-Trityl-2-amino-3-hydroxypropionic acid triethyl ammonium salt

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: N-trityl-L-serine triethyl ammonium salt

OR53186-1G OR53186-25G OR53186-5G
APOSOR53186-1GEA 54.2 EUR
APOSOR53186-1G APOSOR53186-25G APOSOR53186-5G
(S)-Trityl-2-amino-3-hydroxypropionic acid triethyl ammonium salt
(S)-Trityl-2-amino-3-hydroxypropionic acid triethyl ammonium salt
Formule: C₂₂H₂₁NO₃·(C₂H₅)₃N
MW: 448.597 g/mol
Smeltpunt: 144…158 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD01075008
CAS nummer: 111061-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR