4-Bromo-2-iodoaniline

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

OR5425-25G OR5425-5G OR5425-1G OR5425-100G
APOSOR5425-25GEA 43.8 EUR
APOSOR5425-25G APOSOR5425-5G APOSOR5425-1G APOSOR5425-100G
4-Bromo-2-iodoaniline
4-Bromo-2-iodoaniline
Formule: C₆H₅BrIN
Smeltpunt: 69…72 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD02093764
CAS nummer: 66416-72-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR