2-Amino-6-bromoquinoline

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

OR54430-1G
APOSOR54430-1GEA 271 EUR
APOSOR54430-1G
2-Amino-6-bromoquinoline
2-Amino-6-bromoquinoline
Formule: C₉H₇BrN₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06738670
CAS nummer: 791626-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR