Benzyl(dimethyl)amine 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: DMBA, N,N-Dimethylbenzylamine, N-Benzyldimethylamine, BDMA

OR54497-2.5L OR54497-500ML
APOSOR54497-2.5LEA 139 EUR
APOSOR54497-2.5L APOSOR54497-500ML
Benzyl(dimethyl)amine 99%
Benzyl(dimethyl)amine
Formule: C₆H₅CH₂N(CH₃)₂
Kookpunt: 183…184 °C/0,75mmHg
Smeltpunt: –75 °C
Dichtheid: 0,9 g/cm³ (25 °C)
Vlampunt: 53 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008329
CAS nummer: 103-83-3
UN: 2619
ADR: 8+3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR